Barium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Barium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 g
Packaging:
Ampule
Catalog number 047118.14
also known as 047118-14
Price (USD)/ Each
463.00
-
Add to cart
Quantity:
25 g
Packaging:
Ampule
Request bulk or custom format
Price (USD)/ Each
463.00
Add to cart
Barium iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number047118.14
Price (USD)/ Each
463.00
-
Add to cart
Chemical Identifiers
CAS13718-50-8
IUPAC Namebarium(2+) diiodide
Molecular FormulaBaI2
InChI KeySGUXGJPBTNFBAD-UHFFFAOYSA-L
SMILES[I-].[I-].[Ba++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBead
Total Metal Impurities0.001% max.
Particle Size-10 Mesh
Barium iodide is used as a reagent for facile synthesis of poly(phenylcarbyne). It reacts with alkyl potassium compounds to form organobarium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only