Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal Bottles, Thermo Scientific Chemicals
Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal Bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal Bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 047122.AE
also known as 047122-AE
Price (USD)/ Each
44.65
Online exclusive
49.20 
Save 4.55 (9%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
44.65
Online exclusive
49.20 
Save 4.55 (9%)
Add to cart
Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal Bottles, Thermo Scientific Chemicals
Catalog number047122.AE
Price (USD)/ Each
44.65
Online exclusive
49.20 
Save 4.55 (9%)
-
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)50 ppm, max
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)99.80% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only