Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal bottles, Thermo Scientific Chemicals

Catalog number: 047122.K2
1 L, Each
Thermo Scientific Chemicals

Tetrahydrofuran, anhydrous, 99.8+%, BHT-free, over molecular sieves, packaged under Argon in resealable ChemSeal bottles, Thermo Scientific Chemicals

Catalog number: 047122.K2
1 L, Each
Quantity
Catalog number: 047122.K2
also known as 047122-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-99-9
IUPAC Name
oxolane
Molecular Formula
C4H8O
InChI Key
WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES
C1CCOC1
Water Content (Karl Fischer Titration)
50 ppm, max
Appearance (Color)
Clear, colorless
Form
Liquid
Assay (GC)
99.80% min

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text