Sodium oxalate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Sodium oxalate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium oxalate, 0.1N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Catalog number 047123.K2
also known as 047123-K2
Price (USD)/ Each
68.65
Online exclusive
76.10 
Save 7.45 (10%)
-
Add to cart
Request bulk or custom format
Price (USD)/ Each
68.65
Online exclusive
76.10 
Save 7.45 (10%)
Add to cart
Sodium oxalate, 0.1N Standardized Solution, Thermo Scientific Chemicals
Catalog number047123.K2
Price (USD)/ Each
68.65
Online exclusive
76.10 
Save 7.45 (10%)
-
Add to cart
Chemical Identifiers
CAS62-76-0
IUPAC Namedisodium oxalate
Molecular FormulaC2Na2O4
InChI KeyZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA0.0499 M - 0.0501 M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only