Sodium hydroxide, 3.0N Standardized Solution, Thermo Scientific Chemicals
Sodium hydroxide, 3.0N Standardized Solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, 3.0N Standardized Solution, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
Catalog number 047124.K2
also known as 047124-K2
Price (USD)/ Each
54.65
Online exclusive
60.60 
Save 5.95 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
60.60 
Save 5.95 (10%)
Add to cart
Sodium hydroxide, 3.0N Standardized Solution, Thermo Scientific Chemicals
Catalog number047124.K2
Price (USD)/ Each
54.65
Online exclusive
60.60 
Save 5.95 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA2.994 N - 3.006 N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only