Thermo Scientific Chemicals

Toluene, anhydrous, 99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 047136.K2
1 L, Each
Thermo Scientific Chemicals

Toluene, anhydrous, 99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 047136.K2
1 L, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 047136.K2
also known as 047136-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-88-3
IUPAC Name
toluene
Molecular Formula
C7H8
InChI Key
YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1
Form
Liquid
Water Content (Karl Fischer Titration)
≤20 ppm
Appearance (Color)
Clear colorless
Assay from Supplier's CofA
≥99.8%
Residue on Evaporation
≤5 ppm

Description

Toluene is added to gasoline, used to produce benzene, and used as a solvent. It is used in aviation and automotive fuels, as a solvent, and to make other chemicals. Toluene has also been used as a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text