Thermo Scientific Chemicals

Iron(II,III) oxide, nanopowder, 97%, Thermo Scientific Chemicals

Catalog number: 047141.22
100 g, Each
Thermo Scientific Chemicals

Iron(II,III) oxide, nanopowder, 97%, Thermo Scientific Chemicals

Catalog number: 047141.22
100 g, Each
Quantity
Catalog number: 047141.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1317-61-9
IUPAC Name
diiron(3+) λ²-iron(2+) tetraoxidandiide
Molecular Formula
Fe3O4
InChI Key
WTFXARWRTYJXII-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
Appearance (Color)
Black
Assay (unspecified)
96.1% min
Particle Size
50-100nm
Form
Powder
pH
6-8

Description

Iron(II,III) oxide is mainly used as a black-pigment-catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning and as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymers, rubber, fillers, building, construction, appliances and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text