Tin bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Tin bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 047142.MD
also known as 047142-MD
Price (USD)/ Each
44.40
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
44.40
Add to cart
Tin bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Catalog number047142.MD
Price (USD)/ Each
44.40
-
Add to cart
Chemical Identifiers
CAS4600096-08-6
SpecificationsSpecification SheetSpecification Sheet
FormFused solid or viscous liquid as melt
Appearance (Color)White to brown
Assay (unspecified)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only