Rubidium fluoride, 99.1% (metals basis), Thermo Scientific Chemicals
Rubidium fluoride, 99.1% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rubidium fluoride, 99.1% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 047144.18
also known as 047144-18
Price (USD)/ Each
215.65
Online exclusive
239.00 
Save 23.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
215.65
Online exclusive
239.00 
Save 23.35 (10%)
Add to cart
Rubidium fluoride, 99.1% (metals basis), Thermo Scientific Chemicals
Catalog number047144.18
Price (USD)/ Each
215.65
Online exclusive
239.00 
Save 23.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13446-74-7
IUPAC Namerubidium(1+) fluoride
Molecular FormulaFRb
InChI KeyAHLATJUETSFVIM-UHFFFAOYSA-M
SMILES[F-].[Rb+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline
Appearance (Color)White
Total Metal Impurities≤ 0.9%
It is used as raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only