Thermo Scientific Chemicals

Water, LC-MS Grade, Thermo Scientific Chemicals

Catalog number: 047146.K2
1 L, Each
Thermo Scientific Chemicals

Water, LC-MS Grade, Thermo Scientific Chemicals

Catalog number: 047146.K2
1 L, Each
Quantity
Catalog number: 047146.K2
also known as 047146-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
Comment
LC gradient elution suitability at 254nm: Passes Test
LCMS
Negative mode (as Chloramphenicol): ≤50ppb
Comment
Fluorescence at emission maximum for impurities (as Quinine base):≤ 1 ppb
Residue on Evaporation
≤1 ppm
LCMS
Positive mode (as Propazine): ≤50ppb

Description

It is used as mobile phase solvent in LC-MS analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text