Thermo Scientific Chemicals

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 047148.06
5 g, Each
Thermo Scientific Chemicals

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 047148.06
5 g, Each
Quantity
Catalog number: 047148.06
also known as 047148-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13450-91-4
IUPAC Name
gallium(3+) triiodide
Molecular Formula
GaI3
InChI Key
DWRNSCDYNYYYHT-UHFFFAOYSA-K
SMILES
[Ga+3].[I-].[I-].[I-]
Form
Bead
Total Metal Impurities
≤0.001%
Appearance (Color)
Yellow to yellow-green
Comment
-10 mesh

Description

It is useful for the preparation of compounds of gallium(I) and gallium(II) and is reported as useful in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text