Iridium black, 99.95% (metals basis), Thermo Scientific Chemicals
Iridium black, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iridium black, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
10 g
Catalog number 047150.ME
also known as 047150-ME
Price (USD)/ Each
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS7439-88-5
IUPAC Nameiridium
Molecular FormulaIr
InChI KeyGKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES[Ir]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormPowder
Assay (special)Iridium (Ir): 93.0-99.0%
Total Metal Impurities≤0.05%
Surface Area≥20m²/g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only