Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number 047166.03
also known as 047166-03
Price (USD)/ Each
101.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
101.00
Add to cart
Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number047166.03
Price (USD)/ Each
101.00
-
Add to cart
Chemical Identifiers
CAS7790-33-2
IUPAC Namemanganese(2+) diiodide
Molecular FormulaI2Mn
InChI KeyQWYFOIJABGVEFP-UHFFFAOYSA-L
SMILES[Mn++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.01% max.
Appearance (Color)Pink
It is often used in the lighting industry and it is a source of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only