Thermo Scientific Chemicals

Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 047166.09
10 g, Each
Thermo Scientific Chemicals

Manganese(II) iodide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 047166.09
10 g, Each
Quantity
Catalog number: 047166.09
also known as 047166-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-33-2
IUPAC Name
manganese(2+) diiodide
Molecular Formula
I2Mn
InChI Key
QWYFOIJABGVEFP-UHFFFAOYSA-L
SMILES
[Mn++].[I-].[I-]
Particle Size
-10 Mesh
Form
Beads
Residual water
< 100 ppm
Total Metal Impurities
0.01% max.
Appearance (Color)
Pink

Description

It is often used in the lighting industry and it is a source of its derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text