Holmium(III) chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals
Holmium(III) chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Holmium(III) chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 047178.14
also known as 047178-14
Price (USD)/ Each
850.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
850.00
Add to cart
Holmium(III) chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals
Catalog number047178.14
Price (USD)/ Each
850.00
-
Add to cart
Chemical Identifiers
CAS10138-62-2
IUPAC Nameholmium(3+) trichloride
Molecular FormulaCl3Ho
InChI KeyPYOOBRULIYNHJR-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormBead
Appearance (Color)White to pink
Particle Size-10 Mesh
Total Metal Impurities0.05% max.
It is mainly used in research. It is used in ceramics, lasers, nuclear industries and in paramagnetic. It is also employed in calibration of optical spectrophotometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only