Thermo Scientific Chemicals
Semiquantitative Standard 1, Specpure™, Thermo Scientific Chemicals
Catalog number: 047180.AE
100 mL, Each
Thermo Scientific Chemicals
Semiquantitative Standard 1, Specpure™, Thermo Scientific Chemicals
Catalog number: 047180.AE
100 mL, Each
Quantity
Catalog number: 047180.AE
also known as 047180-AE
Price (USD)
Have Questions?
Concentration
Ag 10?g/mL ?2%
Concentration
P 10?g/mL ?2%
Concentration
Pb 10?g/mL ?2%
Concentration
Pr 10?g/mL ?2%
Concentration
Rb 10?g/mL ?2%
Description
Semiquantitative Standard 1, Specpure™ is used as a standard for inductively coupled plasma and direct current plasma (DCP) emission spectrometry for the analysis of trace elements in biomedical and environmental samples.