Thermo Scientific Chemicals

Ruthenium(III) chloride, anhydrous, Ru 47.7% min, Thermo Scientific Chemicals

Catalog number: 047182.09
10 g, Each
Thermo Scientific Chemicals

Ruthenium(III) chloride, anhydrous, Ru 47.7% min, Thermo Scientific Chemicals

Catalog number: 047182.09
10 g, Each
Quantity
Catalog number: 047182.09
also known as 047182-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10049-08-8
IUPAC Name
ruthenium(3+) trichloride
Molecular Formula
Cl3Ru
InChI Key
YBCAZPLXEGKKFM-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ru+3]
Assay (special)
≥48.7% (Ruthenium)
Form
Powder
Appearance (Color)
Black

Description

Ruthenium(III) chloride is widely used as a starting material of ruthenium complexes. It acts as a catalyst used in the oxidative cyclization of 1,7-dienes to oxepane diols. It is used in the hydroxylation of tertiary hydrocarbons in combination of periodate or bromate. It is involved as a catalyst in the synthesis of 2,3-pyridinedicarboxylic acid-13C3, 15N, where the unlabelled analog is an inhibitor of glucose synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text