Thermo Scientific Chemicals

n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 047191.K2
1 L, Each
Thermo Scientific Chemicals

n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog number: 047191.K2
1 L, Each
Quantity
Catalog number: 047191.K2
also known as 047191-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
142-82-5
IUPAC Name
heptane
Molecular Formula
C7H16
InChI Key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES
CCCCCCC
Appearance (Color)
Colorless
Form
Liquid
Assay from Supplier's CofA
≥98.5%
Residue on Evaporation
≤5 ppm
Water Content
≤10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text