Thermo Scientific Chemicals

Cerium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 047194.14
25 g, Each
Thermo Scientific Chemicals

Cerium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 047194.14
25 g, Each
Quantity
Catalog number: 047194.14
also known as 047194-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14457-87-5
IUPAC Name
cerium(3+) tribromide
Molecular Formula
Br3Ce
InChI Key
MOOUSOJAOQPDEH-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Ce+3]
Appearance (Color)
White to gray
Form
Bead
Total Metal Impurities
0.01% max.
Particle Size
-10 Mesh

Description

It is used as an additive for chemical reagent. Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text