Thermo Scientific Chemicals

Zirconium hydride, 99%, Thermo Scientific Chemicals

Catalog number: 047203.09
10 g, Each
Thermo Scientific Chemicals

Zirconium hydride, 99%, Thermo Scientific Chemicals

Catalog number: 047203.09
10 g, Each
Quantity
Catalog number: 047203.09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7704-99-6
IUPAC Name
λ²-zirconium(2+) dihydride
Molecular Formula
H2Zr
InChI Key
IUJYJRCMJZIAJY-UHFFFAOYSA-N
SMILES
[H-].[H-].[Zr++]
Form
Powder, -325 mesh
Assay (unspecified)
≥98.5%
Particle Size
4.5 - 6.5 micron APS
Form
Powder, -325 mesh
Assay (unspecified)
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text