Thermo Scientific Chemicals
Iron, plasma standard solution, Specpure™ Fe 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 047206.AE
100 mL, Each
Thermo Scientific Chemicals
Iron, plasma standard solution, Specpure™ Fe 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 047206.AE
100 mL, Each
Quantity
Catalog number: 047206.AE
also known as 047206-AE
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Fe(NO3)3.9H2O