Thermo Scientific Chemicals

Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations), Thermo Scientific Chemicals

Catalog number: 047209.K2
1 L, Each
Thermo Scientific Chemicals

Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations), Thermo Scientific Chemicals

Catalog number: 047209.K2
1 L, Each
Quantity
Catalog number: 047209.K2
also known as 047209-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-84-9
IUPAC Name
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular Formula
C12H22O11
InChI Key
GUBGYTABKSRVRQ-ASMJPISFSA-N
SMILES
OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Appearance (Color)
Colorless
Form
Liquid
Concentration
0.995% to 1.05% (W/V)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text