PdCl(crotyl)Amphos, Thermo Scientific Chemicals
PdCl(crotyl)Amphos, Thermo Scientific Chemicals
Thermo Scientific Chemicals

PdCl(crotyl)Amphos, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number 047216.03
also known as 047216-03
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
Add to cart
PdCl(crotyl)Amphos, Thermo Scientific Chemicals
Catalog number047216.03
Price (USD)/ Each
302.65
Online exclusive
336.00 
Save 33.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialPdCl(crotyl)Amphos
CAS1334497-06-1
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only