This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047221.AP
also known as 047221-AP
Price (USD)/ Each
232.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
274.00
Save 41.35 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
232.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
274.00
Save 41.35 (15%)
Add to cart
Iron, plasma standard solution, Specpure™ Fe 10,000μg/mL, Thermo Scientific Chemicals
Chemical Name or MaterialIron, plasma standard solution
Name NoteFe 10,000μg/mL
Health Hazard 1H290-H314-H335
Quantity500 mL
Health Hazard 2GHS H Statement H314 Causes severe skin burns and eye damage.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.