Thermo Scientific Chemicals

Hematoxylin hydrate, 80% (dry wt.), water <8%, Thermo Scientific Chemicals

Catalog number: 047223.22
100 g, Each
Thermo Scientific Chemicals

Hematoxylin hydrate, 80% (dry wt.), water <8%, Thermo Scientific Chemicals

Catalog number: 047223.22
100 g, Each
Quantity
Catalog number: 047223.22
also known as 047223-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
517-28-2
IUPAC Name
(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Molecular Formula
C16H14O6
InChI Key
WZUVPPKBWHMQCE-WKTCHCBJNA-N
SMILES
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
Solution Test
Solubility in Alcohol: Pass
Appearance (Color)
Cream to pale brown
Form
Powder
Assay from Supplier's CofA
≥ 80% (dry wt)
Water Content (Karl Fischer Titration)
≤8%

Description

Hematoxylin and eosin together make up Hematoxylin and eosin stain, one of the most commonly used stains in histology. Metal-haematein complexes are used to stain cell nuclei prior to examination under a microscope. Structures that stain with iron- or aluminum-haematein are often called basophilic, even though the mechanism of the staining is different from that of staining with basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text