Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 047235.22
100 g, Each
Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 047235.22
100 g, Each
Quantity
Catalog number: 047235.22
also known as 047235-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-34-1
IUPAC Name
calcium carbonate
Molecular Formula
CCaO3
InChI Key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES
[Ca++].[O-]C([O-])=O
Form
Powder
Total Metal Impurities
10 ppm, max

Description

Applied as neutralizing agent, filler, and extender in rubber, plastics, paints, opacifying agent in paper, putty, antacid, Portland cement, sulfur dioxide, removal from stack gases, allurgical flux, analytical chemistry, carbon dioxide generation, and determining halogens in organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text