Thermo Scientific Chemicals

Iron(III) hydroxide, 20% in H{2}O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted, Thermo Scientific Chemicals

Catalog number: 047241.22
100 g, Each
Thermo Scientific Chemicals

Iron(III) hydroxide, 20% in H{2}O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted, Thermo Scientific Chemicals

Catalog number: 047241.22
100 g, Each
Quantity
Catalog number: 047241.22
also known as 047241-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20344-49-4
IUPAC Name
ironoylol
Molecular Formula
FeHO2
InChI Key
AEIXRCIKZIZYPM-UHFFFAOYSA-M
SMILES
O[Fe]=O
Concentration (weight/weight)
20% min

Description

Iron(III) hydroxide is used in cosmetics, tattoo inks and aquarium water treatment. It is used as a pigment and as an adsorbent for removing lead from aquatic media. It acts as a catalyst for proteomics research. It is involved in the removal of toxic selenate and selenite ions from water systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text