Aluminum oxide, alpha-phase, 99.997% (metals basis), Thermo Scientific Chemicals
Aluminum oxide, alpha-phase, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum oxide, alpha-phase, 99.997% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047261.36
also known as 047261-36
Price (USD)/ Each
406.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
478.00 
Save 71.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
406.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
478.00 
Save 71.35 (15%)
Add to cart
Aluminum oxide, alpha-phase, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number047261.36
Price (USD)/ Each
406.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
478.00 
Save 71.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Comment40 Micron APS
Total Metal Impurities0.003% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only