Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 047268.22
also known as 047268-22
Price (USD)/ Each
657.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
657.00
Add to cart
Strontium iodide, anhydrous, 99% (metals basis), Thermo Scientific Chemicals
Catalog number047268.22
Price (USD)/ Each
657.00
-
Add to cart
Specifications
Chemical Name or MaterialStrontium iodide
Melting Point515°C
Name NoteAnhydrous
CAS10476-86-5
ColorBrown to Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only