Specifications
Chemical Name or MaterialStainless Steel
Name NoteType 430-L
Health Hazard 1H351
Health Hazard 2GHS H Statement
H351-H319-H335
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more