Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Melting Point | 11.8°C |
CAS | 123-91-1 |
Boiling Point | 100°C to 102°C |
Quantity | 4 x 1 L |
Molecular Formula | C{4}H{8}O{2} |
Percent Purity | 99.8% |
Unit Size | Each |
Melting Point | 11.8°C |
CAS | 123-91-1 |
Boiling Point | 100°C to 102°C |
Quantity | 100 mL |
Molecular Formula | C{4}H{8}O{2} |
Percent Purity | 99.8% |
Unit Size | Each |
Melting Point | 11.8°C |
CAS | 123-91-1 |
Boiling Point | 100°C to 102°C |
Quantity | 1 L |
Molecular Formula | C{4}H{8}O{2} |
Percent Purity | 99.8% |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
047305.M1 also known as 047305-M1 | Each | 4 x 1 L | Special offer: 299.65 316.65 Online priceSave 17.00 (5%) Ends: 15-Oct-2024 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
047305.AE also known as 047305-AE | Each | 100 mL | Special offer: 55.65 58.65 Online priceSave 3.00 (5%) Ends: 15-Oct-2024 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
047305.K2 also known as 047305-K2 | Each | 1 L | Special offer: 95.65 100.65 Online priceSave 5.00 (4%) Ends: 15-Oct-2024 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|