You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable AcroSeal™ bottles
Element Search
Structure Search
Actual product may vary
Thermo Scientific™
N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable AcroSeal™ bottles
Have Questions?
Change view
Quantity:
1 L
100 mL
4 x 100 mL
4 x 1 L
Try the chem dex
TM
web app
Catalog number
047390.K2
also known as 047390-K2
Price (USD)
/ Each
146.65
Special Offer
Online exclusive
Ends:
15-Oct-2024
172.00
Save 25.35 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
/ Each
146.65
Special Offer
Online exclusive
Ends:
15-Oct-2024
172.00
Save 25.35 (15%)
Add to cart
N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable AcroSeal™ bottles
Catalog number
047390.K2
Price (USD)
/ Each
146.65
Special Offer
Online exclusive
Ends:
15-Oct-2024
172.00
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
CAS
68-12-2
IUPAC Name
N,N-dimethylformamide
Molecular Formula
C3H7NO
InChI Key
ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES
CN(C)C=O
View more
Specifications
Specification Sheet
Appearance (Color)
Colorless
Assay (GC)
≥99.75% (corrected for water)
Water Content (Karl Fischer Titration)
≤50ppm
Residue on Evaporation
≤5ppm
Form
Liquid
WARNING:
Cancer –
www.P65Warnings.ca.gov