Thermo Scientific Chemicals

Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase, Thermo Scientific Chemicals

Catalog number: 049113.09
10 g, Each
Thermo Scientific Chemicals

Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase, Thermo Scientific Chemicals

Catalog number: 049113.09
10 g, Each
Quantity
Catalog number: 049113.09
also known as 049113-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12033-89-5
IUPAC Name
Silicon nitride
Molecular Formula
N4Si3
InChI Key
HQVNEWCFYHHQES-UHFFFAOYSA-N
SMILES
N12[Si]34N5[Si]11N3[Si]25N41
Total Metal Impurities
0.15% max., 90+% α-phase

Description

Silicon nitride bearings are both full ceramic bearings and ceramic hybrid bearings with balls in ceramics and races in steel. Silicon nitride ceramics have good shock resistance compared to other ceramics. Silicon nitride is used for rocker arm pads for lower wear, turbocharger for lower inertia and less engine lag, and in exhaust gas control valves for increased acceleration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text