Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase, Thermo Scientific Chemicals
Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 049113.18
also known as 049113-18
Price (USD)/ Each
70.50
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
70.50
Add to cart
Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase, Thermo Scientific Chemicals
Catalog number049113.18
Price (USD)/ Each
70.50
-
Add to cart
Chemical Identifiers
CAS12033-89-5
IUPAC NameSilicon nitride
Molecular FormulaN4Si3
InChI KeyHQVNEWCFYHHQES-UHFFFAOYSA-N
SMILESN12[Si]34N5[Si]11N3[Si]25N41
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.15% max., 90+% α-phase
Silicon nitride bearings are both full ceramic bearings and ceramic hybrid bearings with balls in ceramics and races in steel. Silicon nitride ceramics have good shock resistance compared to other ceramics. Silicon nitride is used for rocker arm pads for lower wear, turbocharger for lower inertia and less engine lag, and in exhaust gas control valves for increased acceleration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only