Capitol Vial Sterile Water Testing Vials
Actual product may vary
Thermo Scientific™
Capitol Vial Sterile Water Testing Vials
Capitol Vial Water Testing Vials comply with U.S. EPA requirements for drinking water sample containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Closure TypeDescription
04HP83HCNP118 mLChain of Custody, Flip TopEPA vial, high clarity, sterile, with string, no tablet
04HP83HCPL118 mLChain of Custody, Flip TopEPA vial, high clarity, sterile, with string, 10 mg sodium thiosulfate tablet
2 Options
Catalog number 04HP83HCNP
Price (USD)/ Each
88.90
-
Add to cart
Volume (Metric):
118 mL
Closure Type:
Chain of Custody, Flip Top
Description:
EPA vial, high clarity, sterile, with string, no tablet
Price (USD)/ Each
88.90
Add to cart
Capitol Vial Sterile Water Testing Vials
Catalog number04HP83HCNP
Price (USD)/ Each
88.90
-
Add to cart

Capitol Vial Water Testing Vials comply with U.S. EPA requirements for drinking water sample containers and are available with or without a sodium thiosulfate pill for chlorine neutralization.

Features:

  • Sterile, high clarity polypropylene
  • Available with a 10 mg sodium thiosulfate tablet for sample dechlorination, or without a tablet for unchlorinated water sources
  • Molded with a 100 mL (±2.5 mL) fill line and has one inch of headspace above the fill line to facilitate mixing
  • Integrated tamper evident, chain of custody, and locking seals help ensure container security before and after sample collection

Specifications
DescriptionEPA vial, high clarity, sterile, with string, no tablet
Diameter (Metric) Inner43 mm
ColorHigh Clarity
MaterialHigh Clarity Polypropylene
SterilitySterile
Volume (Metric)118 mL
For Use With (Application)Drinking water analysis
No. per Case200
Volume (English)4 oz.
Closure TypeChain of Custody, Flip Top
Height (Metric)95 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSterile Water Testing Vial
Unit SizeEach