Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 053131.30
250 g, Each
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog number: 053131.30
250 g, Each
Quantity
Catalog number: 053131.30
also known as 053131-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-20-0
IUPAC Name
nickel(2+) hexahydrate dichloride
Molecular Formula
Cl2H12NiO6
InChI Key
LAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
Appearance (Color)
Green
Form
Crystalline powder or crystals
Assay from Supplier's CofA
≥99.95% metals basis
Total Metal Impurities
≤0.05% (500 ppm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text