Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Use With (Equipment) | IonPac AS14 Columns |
Type | Eluent Concentrate |
Standard Concentration | 0.025 |
Concentration | 100 X |
Standard Components | Sodium Carbonate, Sodium Bicarbonate, Water |
Quantity | 250 mL |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Quantity | Concentration | Price (USD) | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
053560 | Each 1 | 250 mL | 100 X | 115.00 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|