Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals
Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals

For Use As a Chemical Intermediate
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 056135.14
also known as 056135-14
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
Add to cart
Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals
Catalog number056135.14
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7789-59-5
IUPAC Namephosphoroyl tribromide
Molecular FormulaBr3OP
InChI KeyUXCDUFKZSUBXGM-UHFFFAOYSA-N
SMILESBrP(Br)(Br)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Assay (special)≥ 98%
FormLump
As a chemical intermediate.Phosphorus(V) oxybromide is used as a brominating agent in the synthesis of 2,4,6-tribromopyrimidine and bromoheterocyclic compounds. It is used in the phosphorus flame retardant and phosphorus activators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only