Thermo Scientific Chemicals

Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals

Catalog number: 056135.22
100 g, Each
Thermo Scientific Chemicals

Phosphorus(V) oxybromide, 98% min, Thermo Scientific Chemicals

Catalog number: 056135.22
100 g, Each
Quantity
Catalog number: 056135.22
also known as 056135-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-59-5
IUPAC Name
phosphoroyl tribromide
Molecular Formula
Br3OP
InChI Key
UXCDUFKZSUBXGM-UHFFFAOYSA-N
SMILES
BrP(Br)(Br)=O
Appearance (Color)
White to yellow
Assay (special)
≥ 98%
Form
Lump

Description

As a chemical intermediate.Phosphorus(V) oxybromide is used as a brominating agent in the synthesis of 2,4,6-tribromopyrimidine and bromoheterocyclic compounds. It is used in the phosphorus flame retardant and phosphorus activators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text