Thermo Scientific Chemicals

12-Molybdophosphoric acid hydrate, ACS, Thermo Scientific Chemicals

Catalog number: 056166.36
500 g, Each
Thermo Scientific Chemicals

12-Molybdophosphoric acid hydrate, ACS, Thermo Scientific Chemicals

Catalog number: 056166.36
500 g, Each
Quantity
Catalog number: 056166.36
also known as 056166-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51429-74-4
IUPAC Name
12-Molybdophosphoric acid
Molecular Formula
H3Mo12O40P
Molecular Weight (g/mol)
1825.40
Synonym
phosphoric acid dodecamolybdenum hexatriacontaoxidandiide
Impurity content
Ca 0.02%
Impurity content
Cl 0.02%
Impurity content
Fe 0.005%
Impurity content
NH₄ 0.01%
Impurity content
SO₄ 0.025%

Description

12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text