Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
High quality sodium acetate eluent and reliable histidine standards are crucial to reliable amino acid analysis. Ensure the best possible results by using Thermo Scientific™ Dionex™ AAA-Direct Reagents with your Dionex™ AAA-Direct system. Reagents include high-grade, contaminant-free sodium acetate and reliable histidine standard are pre-qualified and guaranteed not to contaminate your system. The sodium acetate is preweighed for convenient mixing —all you need to do is add water to produce the correct concentration of eluent.
For Use With (Equipment) | AAA-Direct System |
Type | AAA-Direct Reagents |
Standard Concentration | 100% |
Hazards | Irritant |
Includes | Certificate of Analysis |
Standard Components | Sodium Acetate |
Description | Dionex Sodium Acetate Salt (Pre-weighed Reagent), 2-Pk., Electrochemical-grade |
Storage | Room Temperature |
Unit Size | Each 1 |
For Use With (Equipment) | AAA-Direct System |
Type | AAA-Direct Reagents |
Standard Concentration | 8 μM |
Hazards | Irritant |
Includes | Certificate of Analysis |
Standard Components | L-Histidine HCl |
Description | Dionex Histidine Standard, AAA-Direct System Installation |
Storage | Room Temperature |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
059326 | Each 1 | Dionex Sodium Acetate Salt (Pre-weighed Reagent), 2-Pk., Electrochemical-grade | 76.40 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
059540 | Each 1 | Dionex Histidine Standard, AAA-Direct System Installation | 84.40 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
About the Dionex AAA-Direct System
The Dionex AAA-Direct system is a highly sensitive and direct-detection type of amino acid analyzer that requires no sample derivatization. The Dionex AAA-Direct ion chromatography system uses integrated pulsed amperometry (IPAD)—a type of electrochemical detection—to directly detect amino acids. The Dionex AAA-Direct also provides complete separation of all common amino acids using anion-exchange chromatography (Thermo Scientific™ Dionex™ AminoPac™ PA10 column).
Note: AAA-Direct users are recommended to purchase only the prequalified sodium acetate to guarantee trouble-free use of their system.