Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type |
---|---|
062000 | Salt Tablets, 25 kg |
061000 | Hardness Test Kit |
061160 | Mounting Block w/ Blending Valve 1 in. |
062001 | Bacteria Guard |
063998 | Sensor Controller 2 in. |
063999 | Sensor Controller 1 in. |
063997 | Sensor Controller 1.5 in. |
150111 | Blending Valve, 1 in. |
150110 | Blending Valve, 1.5 in. |
Sensor controller for water softening monitoring
Total hardness measurement (titration solution)
Bacteria guard for water softeners
Flexible connecting hose 1in. with stainless steel jacketing, length 1 meter
Mounting block 1in. with blending valve
Blending valve OVP for 1in. softening systems for permanent hardness adjustment
Blending valve OVP for 1 1/2in. softening systems for permanent hardness adjustment
Share catalog number, name or link