Water Softener Accessories
Water Softener Accessories
Actual product may vary
Thermo Scientific™
Water Softener Accessories
Remove hard ions, such as calcium and magnesium, using these accessories. Softened water extends the life of the reverse osmosis membrane.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
062001Bacteria Guard
061000Hardness Test Kit
061160Mounting Block w/ Blending Valve 1 in.
062000Salt Tablets, 25 kg
063998Sensor Controller 2 in.
063999Sensor Controller 1 in.
063997Sensor Controller 1.5 in.
150111Blending Valve, 1 in.
150110Blending Valve, 1.5 in.
Catalog number 062001
Price (USD)
-
Request A Quote
Type:
Bacteria Guard
Ask our AI about this Product
Remove hard ions, such as calcium and magnesium, using Thermo Scientific™ Water Softener Accessories. Softened water extends the life of the reverse osmosis membrane.
Sensor controller for water softening monitoring
  • The Sensor controller SE is a monitoring system for a quality-controlled operation of the water softening system
Total hardness measurement (titration solution)
  • Necessary to determine the feed water hardness and calculate the softener program settings
Bacteria guard for water softeners
  • For disinfecting the ion exchange bed during each regeneration
  • The bacteria guard extracts chlorine from a part of the salt water to disinfect the ion exchange resin
Flexible connecting hose 1in. with stainless steel jacketing, length 1 meter
  • Smart and easy connection of your Thermo Scientific Water softener to wall connectors
  • Alternative instead of fixed piping, which offers more flexibility for maintenance
Mounting block 1in. with blending valve
  • For comfortable bypassing of the softener for maintenance, reparations, etc
Blending valve OVP for 1in. softening systems for permanent hardness adjustment
  • If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Blending valve OVP for 1 1/2in. softening systems for permanent hardness adjustment
  • If a specific water hardness is required after the softener, the blending valve will some hard water into the soft water, coming out of the softeners output
Specifications
For Use With (Equipment)Double Softening System WMS
TypeBacteria Guard
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection