Thermo Scientific Chemicals

Sodium sulfide, anhydrous, Thermo Scientific Chemicals

Catalog number: 065122.22
100 g, Each
Thermo Scientific Chemicals

Sodium sulfide, anhydrous, Thermo Scientific Chemicals

Catalog number: 065122.22
100 g, Each
Quantity
Catalog number: 065122.22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1313-82-2
IUPAC Name
disodium sulfanediide
Molecular Formula
Na2S
InChI Key
GRVFOGOEDUUMBP-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[S--]
Assay (unspecified)
95% min

Description

In organic chemicals, dyes rayon, leather and paper, in hydrometallurgy of gold ore, sulfiding oxidized lead and copper ores, and as an analytical reagentSodium sulfide is used mainly in the pulp and paper industry in the Kraft process. It serves as an oxygen scavenger agent in water treatment. Other applications include ore flotation, oil recovery, making dyes and detergents. In the leather processing industry, it is utilized for liming operation as an unhairing agent. It is also used in the textile industry as a bleaching and desulfurizing agent and as a dechlorinating agent in chemical photography for toning black and white photographs as well as metals precipitant. It plays an important role as a sulfonation and sulfomethylation agent in the chemical industry and in the production of rubber chemicals, sulfur dyes and other chemical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text