Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 068107.09
also known as 068107-09
Price (USD)/ Each
85.65
Online exclusive
95.50 
Save 9.85 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
95.50 
Save 9.85 (10%)
Add to cart
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Catalog number068107.09
Price (USD)/ Each
85.65
Online exclusive
95.50 
Save 9.85 (10%)
-
Add to cart
Chemical Identifiers
CAS3425-46-5
IUPAC Namepotassium cyanoselanide
Molecular FormulaCKNSe
InChI KeyKYEKHFSRAXRJBR-UHFFFAOYSA-M
SMILES[K+].[Se-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Potassium selenocyanate is used to prepare triphenylphosphine selenide by reacting with triphenylphosphine. It finds application in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury thiocyanate and selenocyanate compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only