Thermo Scientific Chemicals

Dimethyl selenide, Thermo Scientific Chemicals

Catalog number: 068116.03
1 g, Each
Thermo Scientific Chemicals

Dimethyl selenide, Thermo Scientific Chemicals

Catalog number: 068116.03
1 g, Each
Quantity
Catalog number: 068116.03
also known as 068116-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
593-79-3
IUPAC Name
(methylselanyl)methane
Molecular Formula
C2H6Se
InChI Key
RVIXKDRPFPUUOO-UHFFFAOYSA-N
SMILES
C[Se]C
Assay (GC)
> 97.0%
Form
Clear Liquid

Description

Used as primary and secondary intermediate and also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text