Dimethyl diselenide, 99%, Thermo Scientific Chemicals
Dimethyl diselenide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl diselenide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 068119.03
also known as 068119-03
Price (USD)/ Each
92.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
92.70
Add to cart
Dimethyl diselenide, 99%, Thermo Scientific Chemicals
Catalog number068119.03
Price (USD)/ Each
92.70
-
Add to cart
Chemical Identifiers
CAS7101-31-7
IUPAC Name(methyldiselanyl)methane
Molecular FormulaC2H6Se2
InChI KeyVLXBWPOEOIIREY-UHFFFAOYSA-N
SMILESC[Se][Se]C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow-red or orange to very dark orange or red-orange
FormLiquid
Assay (GC)≥98.5%
Dimethyl diselenide is a powerful reagent used for identification of distonic radical cations

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only