Thermo Scientific Chemicals

Silicon(IV) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 069182.K2
1 L, Each
Thermo Scientific Chemicals

Silicon(IV) chloride, 99%, Thermo Scientific Chemicals

Catalog number: 069182.K2
1 L, Each
Quantity
Catalog number: 069182.K2
also known as 069182-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-04-7
IUPAC Name
tetrachlorosilane
Molecular Formula
Cl4Si
InChI Key
FDNAPBUWERUEDA-UHFFFAOYSA-N
SMILES
Cl[Si](Cl)(Cl)Cl
Form
Liquid
Assay from Suppliers CofA
≥98.5%
Impurity content
Report impurities

Description

Silicon(IV) chloride is used as an intermediate in the manufacture of polysilicon. It is also used in the semiconductors industry, photovoltaic cells and in the production of optical fibers. Its derivatives like trichlorosilane and silane are used in the Siemens process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text