Thermo Scientific Chemicals

Tin(II) iodide, 99+%, Thermo Scientific Chemicals

Catalog number: 071112.09
10 g, Each
Thermo Scientific Chemicals

Tin(II) iodide, 99+%, Thermo Scientific Chemicals

Catalog number: 071112.09
10 g, Each
Quantity
Catalog number: 071112.09
also known as 071112-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10294-70-9
IUPAC Name
λ²-tin(2+) diiodide
Molecular Formula
I2Sn
InChI Key
JTDNNCYXCFHBGG-UHFFFAOYSA-L
SMILES
[Sn++].[I-].[I-]
Form
Powder, crystals or crystalline powder
Assay from Supplier's CofA
≥99.0%
Appearance (Color)
Red to orange

Description

Tin(II) iodide is used to prepare organic-inorganic perovskite thin films which exhibit uniform surfaces and strong photoluminescence. It finds application as an analytical reagent in the determination of rhodium and iridium by spectrophotometry. It is also involved in the synthesis of CH3(CH2)11NH3]SnI3, which is a hybrid semiconductor with MoO3 type tin(II) iodide layers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text