Thermo Scientific Chemicals

Di-n-butyltin bis(2-ethylhexanoate), Thermo Scientific Chemicals

Catalog number: 071126.14
25 g, Each
Thermo Scientific Chemicals

Di-n-butyltin bis(2-ethylhexanoate), Thermo Scientific Chemicals

Catalog number: 071126.14
25 g, Each
Quantity
Catalog number: 071126.14
also known as 071126-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2781-10-4
IUPAC Name
dibutyl[(2-ethylhexanoyl)oxy]stannyl 2-ethylhexanoate
Molecular Formula
C24H48O4Sn
InChI Key
GPDWNEFHGANACG-UHFFFAOYNA-L
SMILES
CCCCC(CC)C(=O)O[Sn](CCCC)(CCCC)OC(=O)C(CC)CCCC
Appearance (Color)
White to off-white
Form
Solid
Elemental Analysis
Sn: 20.0% to 25.8%

Description

Di-n-butyltin bis(2-ethylhexanoate) is used in silanol condensation reactions for caulk and sealant applications and in the production of silicones. It is also used in one or two component systems which are compatible with aromatic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text